Ethyl-5-amino-1,3,4-oxadiazole-2-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 5-amino-1,3,4-oxadiazole-2-carboxylate

A734103-1G A734103-25G A734103-5G
76776-114EA 19.1 USD
76776-114 76776-116 76776-118
Ethyl-5-amino-1,3,4-oxadiazole-2-carboxylate 98%
Ethyl-5-amino-1,3,4-oxadiazole-2-carboxylate

Precaución: for Research Use Only

Formula: C₅H₇N₃O₃
MW: 157.13 g/mol
MDL Number: MFCD03425200
CAS Number: 4970-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR