N-Ethyl-N-(3-sulfopropyl)-m-anisidine sodium 97%

Supplier: Ambeed
Warning

Synonyms: N-Ethyl-N-(3-sulphopropyl)-m-anisidine sodium salt

A733050-100MG A733050-1G A733050-1MG A733050-250MG A733050-25G A733050-5G
76776-100EA 7 USD
76776-100 76776-102 76776-104 76776-106 76776-108 76776-110
N-Ethyl-N-(3-sulfopropyl)-m-anisidine sodium 97%
N-Ethyl-N-(3-sulphopropyl)-m-anisidine sodium

Precaución: for Research Use Only

Formula: C₁₂H₁₈NNaO₄S
MDL Number: MFCD00083320
CAS Number: 82611-88-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR