5-Bromo-6-chloro-3-indolyl phosphate disodium salt 95%

Supplier: Ambeed
Warning

Synonyms: Magenta-phos

A725922-1G A725922-250MG
76776-090EA 157.02 USD
76776-090 76776-092
5-Bromo-6-chloro-3-indolyl phosphate disodium salt 95%
5-Bromo-6-chloro-3-indolyl phosphate disodium salt

Precaución: for Research Use Only

Formula: C₈H₄BrClNO₄P·2Na
MW: 370.432 g/mol
MDL Number: MFCD01311741
CAS Number: 404366-59-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR