4-tert-Butoxybenzonitrile 95%

Supplier: Ambeed
Warning

A724266-25G A724266-5G
76776-084EA 155 USD
76776-084 76776-086
4-tert-Butoxybenzonitrile 95%
4-tert-Butoxybenzonitrile

Precaución: for Research Use Only

Formula: C₁₁H₁₃NO
MDL Number: MFCD03425273
CAS Number: 185259-36-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR