6-Methyl-1,2,3,4-tetrahydroquinoline 98%

Supplier: Ambeed
Warning

Synonyms: 1,2,3,4-Tetrahydro-6-methylquinoline

A721576-1G A721576-25G A721576-5G
76776-078EA 22.28 USD
76776-078 76776-080 76776-082
6-Methyl-1,2,3,4-tetrahydroquinoline 98%
6-Methyl-1,2,3,4-tetrahydroquinoline

Precaución: for Research Use Only

Formula: C₁₀H₁₃N
MW: 147.22 g/mol
MDL Number: MFCD00023887
CAS Number: 91-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR