Amidinothiourea 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Guanylthiourea
A721552-1000G
A721552-100G
A721552-10G
A721552-25G
A721552-500G
76776-068EA
211.12
USD
76776-068
76776-070
76776-072
76776-074
76776-076
Amidinothiourea 98%
Amidinothiourea
Precaución: for Research Use Only
Formula:
C₂H₆N₄S MW: 118.158 g/mol |
MDL Number:
MFCD00014472 CAS Number: 2114-02-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...