5-Bromo-4-chloro-3-indolyl phosphate disodium salt (BCIP-Na) 97%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-4-chloro-1H-indol-3-yl phosphate sodium, X-phosphate disodium salt, Sodium 5-bromo-4-chloro-1H-indol-3-yl phosphate

A719887-100MG A719887-1G A719887-250MG A719887-5G
76776-052EA 30.77 USD
76776-052 76776-054 76776-056 77627-466
5-Bromo-4-chloro-3-indolyl phosphate disodium salt (BCIP-Na) 97%
5-Bromo-4-chloro-3-indolyl phosphate disodium salt (BCIP-Na)

Precaución: for Research Use Only

Formula: C₈H₄BrClNNa₂O₄P
MW: 370.43 g/mol
MDL Number: MFCD00036757
CAS Number: 102185-33-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR