5-Methyl-2-(trifluoromethyl)-3-furoic acid 97%

Supplier: Ambeed
Danger

Synonyms: 5-Methyl-2-(trifluoromethyl)furan-3-carboxylic acid

A717451-10G A717451-1G A717451-250MG A717451-5G
76776-036EA 248.26 USD
76776-036 76776-038 76776-040 76776-042
5-Methyl-2-(trifluoromethyl)-3-furoic acid 97%
5-Methyl-2-(trifluoromethyl)-3-furoic acid

Precaución: for Research Use Only

Formula: C₇H₅F₃O₃
MW: 194.108 g/mol
MDL Number: MFCD00275563
CAS Number: 17515-74-1
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR