2-Chloro-5-fluorotoluene 98%

Supplier: Ambeed
Danger

A716859-25G A716859-5G
76776-032EA 17 USD
76776-032 76776-034
2-Chloro-5-fluorotoluene 98%
2-Chloro-5-fluorotoluene

Precaución: for Research Use Only

Formula: C₇H₆ClF
MDL Number: MFCD00040853
CAS Number: 33406-96-1
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR