5-(Acetoacetamido)-2-benzimidazolinone 98%

Supplier: Ambeed
Warning

Synonyms: 5-Acetoacetlamino benzimdazolone

A716680-1000G A716680-100G A716680-25G A716680-500G
76776-024EA 196.27 USD
76776-024 76776-026 76776-028 76776-030
5-(Acetoacetamido)-2-benzimidazolinone 98%
5-(Acetoacetamido)-2-benzimidazolinone

Precaución: for Research Use Only

Formula: C₁₁H₉N₃O₃
MW: 231.21 g/mol
MDL Number: MFCD00444508
CAS Number: 26576-46-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR