3-Bromo-2,6-diluorotoluene 98%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-2,6-difluorotoluene, 1-Bromo-2,4-difluoro-3-methylbenzene

A716326-1G A716326-25G A716326-5G
76776-018EA 8.49 USD
76776-018 76776-020 76776-022
3-Bromo-2,6-diluorotoluene 98%
3-Bromo-2,6-diluorotoluene

Precaución: for Research Use Only

Formula: C₇H₅BrF₂
MW: 207.02 g/mol
MDL Number: MFCD03412221
CAS Number: 221220-97-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR