2-Amino-4-phenylphenol 98%

Supplier: Ambeed
Warning

Synonyms: 3-Amino-4-hydroxybiphenyl, 3-Amino-[1,1'-biphenyl]-4-ol

A715315-1G A715315-5G
76776-004EA 25.47 USD
76776-004 76776-006
2-Amino-4-phenylphenol 98%
2-Amino-4-phenylphenol

Precaución: for Research Use Only

Formula: C₁₂H₁₁NO
MW: 185.2200 g/mol
MDL Number: MFCD00059187
CAS Number: 1134-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR