7-Hydroxy-4-methylcoumarin-3-acetic acid 98%

Supplier: Ambeed
Warning

A711116-250MG A711116-1G
76775-982EA 63 USD
76775-982 77878-553
7-Hydroxy-4-methylcoumarin-3-acetic acid 98%
7-Hydroxy-4-methylcoumarin-3-acetic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₀O₅
MDL Number: MFCD00467589
CAS Number: 5852-10-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR