4-Chloro-5-(chlorosulfonyl)-2-fluorobenzoic acid 97%

Supplier: Ambeed
Danger

A710991-10G A710991-25G A710991-5G
76775-976EA 135.8 USD
76775-976 76775-978 76775-980
4-Chloro-5-(chlorosulfonyl)-2-fluorobenzoic acid 97%
4-Chloro-5-(chlorosulfonyl)-2-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃Cl₂FO₄S
MW: 273.06 g/mol
MDL Number: MFCD06658273
CAS Number: 56447-54-2
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR