5-Chloro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene 98%

Supplier: Ambeed
Warning

A710528-250MG A710528-25G
76775-974EA 8 USD
76775-974 77878-552
5-Chloro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene 98%
5-Chloro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene

Precaución: for Research Use Only

Formula: C₁₀H₁₄BClO₂S
MDL Number: MFCD08063140
CAS Number: 635305-24-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR