8-Amino-5-bromoisoquinoline 98%

Supplier: Ambeed
Danger

Synonyms: 5-Bromoisoquinolin-8-amine

A710122-1G A710122-250MG A710122-5G
76775-968EA 40.32 USD
76775-968 76775-970 76775-972
8-Amino-5-bromoisoquinoline 98%
8-Amino-5-bromoisoquinoline

Precaución: for Research Use Only

Formula: C₉H₇BrN₂
MW: 223.0693 g/mol
MDL Number: MFCD09261047
CAS Number: 90721-35-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR