(4-(1H-1,2,4-Triazol-1-yl)phenyl)methanol 97%

Supplier: Ambeed
Warning

Synonyms: 4-(1,2,4-Triazol-1-yl)benzyl Alcohol

A709150-1G A709150-5G
76775-964EA 28.65 USD
76775-964 76775-966
(4-(1H-1,2,4-Triazol-1-yl)phenyl)methanol 97%
(4-(1H-1,2,4-Triazol-1-yl)phenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₉N₃O
MW: 175.1873 g/mol
MDL Number: MFCD02682061
CAS Number: 143426-50-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR