2-Chloro-5-(trifluoromethyl)benzene-1,3-diamine 95%

Supplier: Ambeed
Warning

A709126-100MG A709126-1G A709126-250MG
76775-958EA 27.59 USD
76775-958 76775-960 76775-962
2-Chloro-5-(trifluoromethyl)benzene-1,3-diamine 95%
2-Chloro-5-(trifluoromethyl)benzene-1,3-diamine

Precaución: for Research Use Only

Formula: C₇H₆ClF₃N₂
MW: 210.58 g/mol
MDL Number: MFCD00466370
CAS Number: 34207-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR