6-Amino-2,3-difluorophenol 95%

Supplier: Ambeed
Warning

A707456-100MG A707456-1G A707456-250MG A707456-5G A707456-25G
76775-950EA 15 USD
76775-950 76775-952 76775-954 76775-956 77878-547
6-Amino-2,3-difluorophenol 95%
6-Amino-2,3-difluorophenol

Precaución: for Research Use Only

Formula: C₆H₅F₂NO
MDL Number: MFCD03094310
CAS Number: 115551-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR