5-Chloro-3-methyl-1-benzothiophene-2-carboxylic acid 90%

Supplier: Ambeed
Warning

Synonyms: 5-Chloro-3-methylbenzo[b]thiophene-2-carboxylic acid

A707194-1G A707194-250MG A707194-100MG
76775-946EA 138.98 USD
76775-946 76775-948 77627-446
5-Chloro-3-methyl-1-benzothiophene-2-carboxylic acid 90%
5-Chloro-3-methyl-1-benzothiophene-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₇ClO₂S
MW: 226.68 g/mol
MDL Number: MFCD03407361
CAS Number: 50451-84-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR