(-)-Nopol 98%

Supplier: Ambeed
Warning

A705693-100G A705693-500G
76775-940EA 49.87 USD
76775-940 76775-942
(-)-Nopol 98%
(-)-Nopol

Precaución: for Research Use Only

Formula: C₁₁H₁₈O
MW: 166.26 g/mol
MDL Number: MFCD00075187
CAS Number: 35836-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR