5-Phenylvaleric acid 99%

Supplier: Ambeed
Warning

Synonyms: 5-Phenylpentanoic acid, 5-Phenylvaleric Acid

A705523-100G A705523-10G A705523-1G A705523-25G A705523-5G
76775-930EA 411.63 USD
76775-930 76775-932 76775-934 76775-936 76775-938
5-Phenylvaleric acid 99%
5-Phenylvaleric acid

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
MDL Number: MFCD00004416
CAS Number: 2270-20-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR