3-[1-(Dimethylamino)ethyl]phenol 97%

Supplier: Ambeed
Danger

Synonyms: 3-(1-(Dimethylamino)ethyl)phenol

A704153-10G A704153-1G A704153-25G A704153-5G
76775-922EA 37.14 USD
76775-922 76775-924 76775-926 76775-928
3-[1-(Dimethylamino)ethyl]phenol 97%
3-[1-(Dimethylamino)ethyl]phenol

Precaución: for Research Use Only

Formula: C₁₀H₁₅NO
MW: 165.236 g/mol
MDL Number: MFCD08272688
CAS Number: 105601-04-5
UN: 2923
ADR: 8,6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR