2-Chloro-4,5-difluorobenzonitrile 98%

Supplier: Ambeed
Warning

A698887-1G A698887-25G A698887-5G
76775-898EA 17 USD
76775-898 77627-416 77627-418
2-Chloro-4,5-difluorobenzonitrile 98%
2-Chloro-4,5-difluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₂ClF₂N
MDL Number: MFCD02871619
CAS Number: 135748-34-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR