5-(4-Methoxyphenyl)isoxazole 95%

Supplier: Ambeed
Warning

A697570-1G A697570-250MG A697570-5G
76775-888EA 77 USD
76775-888 76775-890 76775-892
5-(4-Methoxyphenyl)isoxazole 95%
5-(4-Methoxyphenyl)isoxazole

Precaución: for Research Use Only

Formula: C₁₀H₉NO₂
MDL Number: MFCD02183545
CAS Number: 3672-48-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR