1-Bromo-2,3,4,5-tetrafluorobenzene 98%

Supplier: Ambeed
Danger

A697379-250MG A697379-5G A697379-1G
76775-884EA 7.43 USD
76775-884 76775-886 77627-408
1-Bromo-2,3,4,5-tetrafluorobenzene 98%
1-Bromo-2,3,4,5-tetrafluorobenzene

Precaución: for Research Use Only

Formula: C₆HBrF₄
MW: 228.97 g/mol
MDL Number: MFCD00017900
CAS Number: 1074-91-5
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR