6-Bromo-2,3-difluorobenzyl alcohol 97%

Supplier: Ambeed
Warning

A695775-1G A695775-250MG A695775-5G
76775-878EA 25 USD
76775-878 76775-880 76775-882
6-Bromo-2,3-difluorobenzyl alcohol 97%
6-Bromo-2,3-difluorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₅BrF₂O
MDL Number: MFCD07368109
CAS Number: 651326-72-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR