6-Bromo-8-fluoronaphthalen-2-ol 95%

Supplier: Ambeed
Warning

A695506-100MG A695506-1G A695506-250MG
76775-864EA 131.56 USD
76775-864 76775-866 76775-868
6-Bromo-8-fluoronaphthalen-2-ol 95%
6-Bromo-8-fluoronaphthalen-2-ol

Precaución: for Research Use Only

Formula: C₁₀H₆BrFO
MW: 241.05 g/mol
MDL Number: MFCD09037977
CAS Number: 82995-06-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR