5-Bromo-α,α,α,2-tetrafluorotoluene 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-2-fluorobenzotrifluoride, 1-Bromo-4-fluoro-3-(trifluoromethyl)benzene

A693258-100G A693258-10G A693258-25G A693258-500G A693258-5G
76775-848EA 48 USD
76775-848 76775-850 76775-852 76775-854 76775-856
5-Bromo-α,α,α,2-tetrafluorotoluene 98%
5-Bromo-α,α,α,2-tetrafluorotoluene

Precaución: for Research Use Only

Formula: C₇H₃BrF₄
MDL Number: MFCD00042493
CAS Number: 393-37-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR