4-(2-Aminoethyl)benzenesulfonamide 98%

Supplier: Ambeed
Danger

A690542-1000G A690542-100G A690542-10G A690542-25G A690542-500G A690542-5G
76775-832EA 264.17 USD
76775-832 76775-834 76775-836 76775-838 76775-840 76775-842
4-(2-Aminoethyl)benzenesulfonamide 98%
4-(2-Aminoethyl)benzenesulphonamide

Precaución: for Research Use Only

Formula: C₈H₁₂N₂O₂S
MW: 200.26 g/mol
MDL Number: MFCD00010301
CAS Number: 35303-76-5
UN: 3259
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR