4-Boronobenzenesulfonamide pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzenesulphonamide, 4-Sulphamoylbenzeneboronic acid, pinacol ester

A690264-100MG A690264-1G A690264-250MG A690264-5G
76775-824EA 26.53 USD
76775-824 76775-826 76775-828 76775-830
4-Boronobenzenesulfonamide pinacol ester 98%
4-Boronobenzenesulphonamide pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₈BNO₄S
MW: 283.1516 g/mol
MDL Number: MFCD06657806
CAS Number: 214360-51-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR