4-[(2,2-Dimethylpropanoyl)amino]benzeneboronic acid 96%

Supplier: Ambeed
Warning

Synonyms: 4-(Tert-butylcarbonylamino)phenylboronic acid, 4-Pivalamidobenzeneboronic acid

A688821-100MG A688821-1G A688821-250MG
76775-810EA 37.14 USD
76775-810 76775-812 76775-814
4-[(2,2-Dimethylpropanoyl)amino]benzeneboronic acid 96%
4-[(2,2-Dimethylpropanoyl)amino]benzeneboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₆BNO₃
MW: 221.061 g/mol
MDL Number: MFCD08235071
CAS Number: 182344-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR