6-Chloroisatoic anhydride 98%

Supplier: Ambeed
Warning

Synonyms: 5-Chloro-2H-3,1-benzoxazine-2,4(1H)-dione

A688302-1G A688302-250MG A688302-5G
76775-794EA 37 USD
76775-794 76775-796 76775-798
6-Chloroisatoic anhydride 98%
6-Chloroisatoic anhydride

Precaución: for Research Use Only

Formula: C₈H₄ClNO₃
MDL Number: MFCD09032504
CAS Number: 20829-96-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR