(4-tert-Butylphenyl)methanethiol 98%

Supplier: Ambeed
Danger

Synonyms: 4-tert-Butylbenzylthiol, (4-(tert-Butyl)phenyl)methanethiol

A687927-25G A687927-100G
76775-792EA 9.55 USD
76775-792 77627-394
(4-tert-Butylphenyl)methanethiol 98%
(4-tert-Butylphenyl)methanethiol

Precaución: for Research Use Only

Formula: C₁₁H₁₆S
MW: 180.31 g/mol
MDL Number: MFCD00068220
CAS Number: 49543-63-7
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR