5-Chloro-2-iodobenzotrifluoride 98%

Supplier: Ambeed
Warning

A687129-100G A687129-25G A687129-500G A687129-5G
76775-784EA 66 USD
76775-784 76775-786 76775-788 76775-790
5-Chloro-2-iodobenzotrifluoride 98%
5-Chloro-2-iodobenzotrifluoride

Precaución: for Research Use Only

Formula: C₇H₃ClF₃I
MDL Number: MFCD00042579
CAS Number: 23399-77-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR