4-Sulfo-1,8-naphthalic anhydride potassium salt 95%

Supplier: Ambeed
Warning

A685871-100G A685871-10G A685871-25G A685871-500G A685871-5G
76775-774EA 47 USD
76775-774 76775-776 76775-778 76775-780 76775-782
4-Sulfo-1,8-naphthalic anhydride potassium salt 95%
4-Sulfo-1,8-naphthalic anhydride potassium salt

Precaución: for Research Use Only

Formula: C₁₂H₆O₆S
MDL Number: MFCD00012097
CAS Number: 71501-16-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR