Ethyl-2-amino-4,5-dimethoxybenzoate 98%

Supplier: Ambeed
Warning

A683181-10G A683181-1G A683181-250MG A683181-5G
76775-758EA 74 USD
76775-758 76775-760 76775-762 76775-764
Ethyl-2-amino-4,5-dimethoxybenzoate 98%
Ethyl-2-amino-4,5-dimethoxybenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₅NO₄
MDL Number: MFCD00017323
CAS Number: 20323-74-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR