5-Bromo-2-methylbenzenesulfonyl chloride 97%

Supplier: Ambeed
Danger

A682732-100MG A682732-1G A682732-250MG A682732-25G A682732-5G
76775-748EA 11 USD
76775-748 76775-750 76775-752 76775-754 76775-756
5-Bromo-2-methylbenzenesulfonyl chloride 97%
5-Bromo-2-methylbenzenesulfonyl chloride

Precaución: for Research Use Only

Formula: C₇H₆BrClO₂S
MDL Number: MFCD06655596
CAS Number: 69321-56-8
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR