6-Ethoxynicotinic acid 98%

Supplier: Ambeed
Warning

A678538-100MG A678538-1G A678538-250MG
76775-742EA 22.28 USD
76775-742 76775-744 76775-746
6-Ethoxynicotinic acid 98%
6-Ethoxynicotinic acid

Precaución: for Research Use Only

Formula: C₈H₉NO₃
MW: 167.17 g/mol
MDL Number: MFCD07403514
CAS Number: 97455-65-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR