Ethyl 4-(trifluoromethyl)thiazole-2-carboxylate 95%
Supplier: Ambeed
|
Warning
|
A678085-100MG
A678085-1G
A678085-250MG
76775-730EA
26.53
USD
76775-730
76775-732
76775-734
Ethyl 4-(trifluoromethyl)thiazole-2-carboxylate 95%
Ethyl 4-(trifluoromethyl)thiazole-2-carboxylate
Precaución: for Research Use Only
Formula:
C₇H₆F₃NO₂S MW: 225.18 g/mol |
MDL Number:
MFCD08669898 CAS Number: 79247-86-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...