2-Bromo-5-fluoro-4-methylbenzonitrile 98+%

Supplier: Ambeed
Warning

A677985-100MG A677985-1G A677985-250MG
76775-724EA 10 USD
76775-724 76775-726 76775-728
2-Bromo-5-fluoro-4-methylbenzonitrile 98+%
2-Bromo-5-fluoro-4-methylbenzonitrile

Precaución: for Research Use Only

Formula: C₈H₅BrFN
MDL Number: MFCD07782072
CAS Number: 916792-11-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR