N-(Ethoxycarbonyl)phthalimide 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 1,3-dioxoisoindoline 2-carboxylate, N-Carbethoxyphthalimide

A676930-100G A676930-10G A676930-25G A676930-500G A676930-5G
76775-714EA 40.32 USD
76775-714 76775-716 76775-718 76775-720 76775-722
N-(Ethoxycarbonyl)phthalimide 98%
N-(Ethoxycarbonyl)phthalimide

Precaución: for Research Use Only

Formula: C₁₁H₉NO₄
MW: 219.20 g/mol
MDL Number: MFCD00005893
CAS Number: 22509-74-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR