2-Chloro-4-(trifluoromethyl)aniline 98%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-3-chloro-α,α,α-trifluorotoluene, 2-Chloro-α,α,α-trifluoro-p-toluidine, 4-Amino-3-chlorobenzotrifluoride

A672393-100G A672393-10G A672393-25G A672393-5G
76775-692EA 54 USD
76775-692 76775-694 77573-166 77598-688
2-Chloro-4-(trifluoromethyl)aniline 98%
2-Chloro-4-(trifluoromethyl)aniline

Precaución: for Research Use Only

Formula: C₇H₅ClF₃N
MW: 195.57 g/mol
Boiling Pt: 89
MDL Number: MFCD00042563
CAS Number: 39885-50-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR