6-Chloropyridine-2-carboxamide 98%

Supplier: Ambeed
Warning

Synonyms: 6-Chloropyridine-2-carboxylic acid amide, 2-Carbamoyl-6-chloropyridine, 6-Chloropicolinamide

A669675-100MG A669675-1G A669675-250MG
76775-680EA 35.01 USD
76775-680 76775-682 76775-684
6-Chloropyridine-2-carboxamide 98%
6-Chloropyridine-2-carboxamide

Precaución: for Research Use Only

Formula: C₆H₅ClN₂O
MW: 156.569 g/mol
MDL Number: MFCD00708892
CAS Number: 70593-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR