Ethyl 5-(tert-butyl)-1H-pyrazole-3-carboxylate 98%

Supplier: Ambeed
Warning

A669251-1G A669251-25G A669251-5G
76775-674EA 14.86 USD
76775-674 76775-676 76775-678
Ethyl 5-(tert-butyl)-1H-pyrazole-3-carboxylate 98%
Ethyl 5-(tert-butyl)-1H-pyrazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₆N₂O₂
MW: 196.25 g/mol
MDL Number: MFCD02278306
CAS Number: 83405-70-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR