3,4-Dimethoxy-2-hydroxymethylpyridine 98%

Supplier: Ambeed
Warning

Synonyms: 3,4-Dimethylpicolinyl alcohol, (3,4-Dimethoxypyridin-2-yl)methanol

A663107-1G A663107-25G A663107-5G
76775-642EA 15.92 USD
76775-642 76775-644 76775-646
3,4-Dimethoxy-2-hydroxymethylpyridine 98%
3,4-Dimethoxy-2-hydroxymethylpyridine

Precaución: for Research Use Only

Formula: C₈H₁₁NO₃
MW: 169.178 g/mol
MDL Number: MFCD06795941
CAS Number: 72830-08-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR