3-Cyanophenyl isocyanate 98%

Supplier: Ambeed
Danger

Synonyms: 3-Cyanophenylisocyanate, 3-Isocyanatobenzonitrile

A661978-1G A661978-250MG A661978-25G A661978-5G
76775-624EA 21.22 USD
76775-624 76775-626 76775-628 76775-630
3-Cyanophenyl isocyanate 98%
3-Cyanophenyl isocyanate

Precaución: for Research Use Only

Formula: C₈H₄N₂O
MW: 144.13 g/mol
MDL Number: MFCD00013856
CAS Number: 16413-26-6
UN: 2206
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR