5-Bromo-2,4-dihydroxybenzoic acid monohydrate 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-2,4-dihydroxybenzoic acid hydrate

A659818-1G A659818-5G
76775-620EA 15.92 USD
76775-620 76775-622
5-Bromo-2,4-dihydroxybenzoic acid monohydrate 98%
5-Bromo-2,4-dihydroxybenzoic acid monohydrate

Precaución: for Research Use Only

Formula: C₇H₇BrO₅
MW: 251.032 g/mol
MDL Number: MFCD00149094
CAS Number: 160348-98-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR