SEW-2871 95%

Supplier: Ambeed
Danger

Synonyms: 5-(4-Phenyl-5-(trifluoromethyl)thiophen-2-yl)-3-(3-(trifluoromethyl)phenyl)-1,2,4-oxadiazole, 5-[4-Phenyl-5-(trifluoromethyl)thiophen-2-yl]-3-[3-(trifluoromethyl)phenyl]-1,2,4-oxadiazole, SEW2871

A656270-10MG A656270-1MG A656270-25MG A656270-5MG
76775-592EA 130.5 USD
76775-592 76775-594 76775-596 76775-598
SEW-2871 95%
SEW-2871

Precaución: for Research Use Only

Formula: C₂₀H₁₀F₆N₂OS
MW: 440.4 g/mol
MDL Number: MFCD00096600
CAS Number: 256414-75-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR