5-Bromo-2-fluorobenzeneboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-2-fluorophenylboronic acid, 5-Bromo-2-fluorobenzeneboronic acid

A655688-100G A655688-1G A655688-25G A655688-5G
76775-584EA 362.83 USD
76775-584 76775-586 76775-588 76775-590
5-Bromo-2-fluorobenzeneboronic acid 98%
5-Bromo-2-fluorobenzeneboronic acid

Precaución: for Research Use Only

Formula: C₆H₅BBrFO₂
MW: 218.82 g/mol
MDL Number: MFCD05664307
CAS Number: 112204-57-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR